Learn More
Retsch™ Software EasySieve™
Product Code. 15706592 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Sieve Shakers AS control / AS 200 tap / AS jet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15706592 Sieve Shakers AS control / AS 200 tap / AS jet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15706592 Supplier Retsch™ Supplier No. 326450031
  • ONLINE EXCLUSIVE PRICE
    1060.00 EUR
    851.57 EUR / Pack of 1
    Save 208.43 EUR
    19% Off
Estimated Shipment: 15-01-2027
Retsch™ Software EasySieve™ is software designed for particle size analysis.
  • Automatic registration, evaluation and administration of measurement data
  • Logical design, self-explanatory
  • Measurement protocol in accordance with standards
  • Easy selection and configuration of common analytical scales
  • English and German
  • For automatic control of sieve shakers AS 200 control, AS 300 control, AS 450 control, AS 400 control, AS 200 tap, AS 200 jet
  • For Windows™ 2000 / XP / Vista / 7 / 8
Specifications
Product Type Software
For Use With (Application) Particle Size Analysis
For Use With (Equipment) Sieve Shakers AS control / AS 200 tap / AS jet