Learn More
Syngene™ Dymension 1 Software
GREENER_CHOICE
Product Code. 12837004 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
Gbox-Chemi-XX6/9
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12837004 Gbox-Chemi-XX6/9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12837004 Supplier Syngene™ Supplier No. DYM1
  • ONLINE EXCLUSIVE PRICE
    1915.00 EUR
    1796.11 EUR / Pack of 1
    Save 118.89 EUR
    6% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Syngene™ Dymension 1 Software is a 2D analysis software for gel electrophoresis.
  • Software for the rapid analysis of 2D protein spots
    between samples containing one gel only
  • Synchronised zoom with intensity autoscale
  • Automatic and precise image warping with vector view
  • Background correction and noise filtering
  • Automatic spot detection 3 x 10MB images
  • Spot detected in eight seconds
  • Automatic spot matching
  • Spot detection with user adjustable parameters
  • Spot editing and filtering Spot normalization
Specifications
For Use With (Application) 2d electrophoresis analysis
For Use With (Equipment) Gbox-Chemi-XX6/9