Learn More
Diversey Soft Care Sensitive
Product Code. 17853105
Change view
Click to view available options
Size:
2 x 5 L
6 x 1 L
10 x 0.5 L
Unit Size:
10 L
5 L
6 L
Packaging:
With pump
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize Packaging
17853105 2 x 5 L 10 L -
17893105 6 x 1 L 6 L -
17873105 10 x 0.5 L 5 L With pump
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17853105 Supplier Diversey Supplier No. 101108201
  • ONLINE EXCLUSIVE PRICE
    43.07 EUR
    40.84 EUR / 10 L
    Save 2.23 EUR
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Fragrance-free, dye-free mild hand wash for sensitive skins. Proves a great solution for areas where frequent washing is required and healthy hands are desired.
  • Biodegradable and ethically sourced
  • Skin compatability tested (Patch test)
  • Eco-certified with Nordic Swan Ecolabel
  • Endorsed by Swedish Allergy and Asthma Foundation
  • Safe, quick and effective hand wash helps reduce the spread of germs
  • Effective surfactant system ensures excellent removal of grease and other soils
  • Wetting agents provide even application and an easy rinse
  • Reduced skin irritation and greater skin compatibility with a pH balanced formula
  • Moisturizes hands with softening emollients, maintaining skinhealth for frequent users
  • Rubs easily onto hands and leaves no sticky residue
Specifications
Product Type Gentle Soap
Color Clear
Form Liquid
Disposable Single-use
Latex Free Yes
Size 2 x 5 L