Learn More
Sofosbuvir-d6, MedChemExpress
Description
Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Specifications
Specifications
| Chemical Name or Material | Sofosbuvir-d6 |
| CAS | 1868135-06-1 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C22H23D6FN3O9P |
| Color | White |
| Synonym | PSI-7977-d6 GS-7977-d6 |
| SMILES | O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)=O)O2)(C)F)C=CC(N1)=O |
| Molecular Weight (g/mol) | 535.49 |
| Quantity | 5 mg |
| Show More |