missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trimetaphosphate, TRC

Product Code. 30425193
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425193 10 g
30419735 1 g
30420026 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425193 Supplier Toronto Research Chemicals Supplier No. TRCS67378010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-84-4
Molecular Formula 3[Na] . O9 P3
Molecular Weight (g/mol) 305.88519
Synonym Metaphosphoric acid (H3P3O9), sodium salt, Metaphosphoric Acid Trisodium Salt, Sodium metaphosphate, Cyclic sodium trimetaphosphate, Detoxol, Polyrinsan 5B, STMP, Sodium cyclotriphosphate, Sodium metaphosphate, Sodium phosphate, Sodium trimetaphosphate, Trimetaphosphoric Acid Trisodium Salt, Trisodium Cyclotriphosphate, Trisodium Metaphosphate, Trisodium Tricyclophosphate, Trisodium Trimetaphosphate
IUPAC Name trisodium;2,4,6-trioxido-1,3,5,2λμ,4λμ,6λμ-trioxatriphosphinane 2,4,6-trioxide
SMILES [Na+].[Na+].[Na+].[O-]P1(=O)OP(=O)([O-])OP(=O)([O-])O1

Specifications

Quantity 10 g
InChI Formula InChI=1S/3Na.H3O9P3/c;;;1-10(2)7-11(3,4)9-12(5,6)8-10/h;;;(H,1,2)(H,3,4)(H,5,6)/q3*+1;/p-3
Formula Weight 305.8448
Chemical Name or Material Sodium Trimetaphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.