missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Pentahydrate, Supelco™

Product Code. 15854840
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854840 1 kg 1kg
15844840 500 g 500g
15827480 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15854840 Supplier Supelco Supplier No. 1065161000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7

Specifications

Melting Point 48°C
Quantity 1 kg
MDL Number MFCD00149186
Synonym Sodium thiosulfate pentahydrate, Antichlor, Sodium thiosulfate pentahydrate
Molecular Weight (g/mol) 248.18
Grade Research
Density 1.74 g/cm3 at 20°C
Chemical Name or Material Sodium Thiosulfate Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.