missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.1M/l Solution, Scharlab

Product Code. 11388584
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388584 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11388584 Supplier Scharlab Supplier No. SO07311000
  • ONLINE EXCLUSIVE PRICE
    17.50 EUR
    16.57 EUR / 1 L
    Save 0.93 EUR
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry as a titrant in volumetric analysis and as a reducing agent.

This volumetric solution was checked by means of potentiometric methods using Scharlau's potassium iodate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

  • Molecular Formula: Na2S2O3 • 5H2O
  • Molar Mass: 248.18g/mol
  • CAS: 10102-17-7
  • EIN ECS No.: 231-867-5
  • Factor: 0.999–1.001

Specifications

CAS 10102-17-7
Physical Form Solution
Quantity 1 L
Molecular Formula H10Na2O8S2
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
Molecular Weight (g/mol) 248.172
PubChem CID 61475
ChEBI CHEBI:32150
Formula Weight 248.18g/mol
Concentration or Composition (by Analyte or Components) 0.1mol/L
Density ∼1.004g/cm3 
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.