missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, Pharmpur, Scharlab

Product Code. 11388564
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388564 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11388564 Supplier Scharlab Supplier No. SO0672005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7757-83-7
Quantity 5 kg
Molecular Formula H14Na2O10S
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
IUPAC Name disodium heptahydrate sulfite
Molecular Weight (g/mol) 252.14
Grade Pharmpur
compliance-icons compliance-icons
Product Identifier
  • Sodium sulfite, extra pure, Pharmpur, Ph Eur, BP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H413-May cause long lasting harmful effects to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.