missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 6106-20-3 |
| Color | White |
| pH | 9.6 |
| Physical Form | Solid |
| Quantity | 3 kg |
| MDL Number | 150353 |
| Synonym | sodium sesquicarbonate, snowfine, urao, magadi soda, snowflake crystals, sodium carbonate 3:2, trisodium hydrogendicarbonate, hsdb 769, sodium sesquicarbonate dihydrate, carbonic acid, sodium salt 2:3 |
| Formula Weight | 226.023g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium sesquicarbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Sodium carbonate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?