Learn More
Sodium Picosulfate, MedChemExpress
Product Code. 16488518
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488518 10 mg 10mg
16418366 100 mg 100mg
16414397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16488518 Supplier Medchem Express Supplier No. HYB0544
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Specifications
Chemical Name or Material Sodium Picosulfate
CAS 10040-45-6
Color White
Purity Grade Notes Research
Molecular Formula C18H13NNa2O8S2
Quantity 10 mg
Synonym Sodium Picosulphate
Solubility Information DMSO : 100 mg/mL (207.72 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (207.72 mM)
SMILES O=S(OC1=CC=C(C=C1)C(C2=CC=CC=N2)C3=CC=C(OS(=O)(O[Na])=O)C=C3)(O[Na])=O
Molecular Weight (g/mol) 481.41
Formula Weight 481.41
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism