missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, TRC

Product Code. 30411077
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411077 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411077 Supplier Toronto Research Chemicals Supplier No. TRCS6480001KG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium nitrite
CAS 7632-00-0
Quantity 1 kg
Molecular Formula N O2 . Na
Synonym Sodium nitrite, Nitrous Acid Sodium Salt, Anti-Rust, E 250, Erinitrit, Filmerine, M 138C, Nitrous Acid Soda, SNS 40, Sodium Nitrite, Sodium Nitrite (NaNO2), Synfat 1004
InChI Formula InChI=1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1
SMILES [Na+].[O-]N=O
IUPAC Name sodium;nitrite
Molecular Weight (g/mol) 69
Formula Weight 68.9827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.