Learn More
Sodium Nitrite, For Analysis, EMSURE™, MilliporeSigma™
Product Code. 15824860
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15824860 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15824860 Supplier Supelco Supplier No. 1065490100
 Restricted Item
Estimated Shipment: 23-02-2027
Recommended Storage
+5°C to +30°C.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O
Specifications
Melting Point 280°C (decomposition)
Color White
pH 9 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 100 g
Solubility Information 820g/L
Formula Weight 69
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.1 g/3 (20 °C)
Chemical Name or Material Sodium Nitrite