missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, Supelco™

Product Code. 15854850
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854850 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15854850 Supplier Supelco Supplier No. 1065351000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

Cryst. EMPLURA™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 308°C
CAS 7631-99-4
Quantity 1 kg
MDL Number MFCD00011119
Synonym Sodium nitrate, Nitric acid sodium salt, Sodium nitrate
Molecular Weight (g/mol) 84.99
Grade Research
Density 2.26 g/cm3 at 20°C
Chemical Name or Material Sodium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.