Learn More
Sodium Nitrate, Supelco™
Product Code. 15814860
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814860 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ100 Graded - For Non-regulated Laboratory Applications
Product Code. 15814860 Supplier Supelco Supplier No. 1065460050
 Restricted Item
Estimated Shipment: 26-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
MQ100 Graded - For Non-regulated Laboratory Applications
99.99 Suprapur™
TRUSTED_SUSTAINABILITY
Tekniske data
Melting Point 308°C
CAS 7631-99-4
Quantity 50 g
MDL Number MFCD00011119
Synonym Sodium nitrate, Nitric acid sodium salt, Sodium nitrate
Molecular Weight (g/mol) 84.99
Grade Research
Density 2.26 g/cm3 at 20°C
Chemical Name or Material Sodium Nitrate
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.