missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, ACS Reagent Grade, Scharlab

Product Code. 11328554
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328554 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11328554 Supplier Scharlab Supplier No. SO05011000
  • ONLINE EXCLUSIVE PRICE
    74.50 EUR
    61.99 EUR / 1kg
    Save 12.51 EUR
    16% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, an oxidizing agent, in the food industry (E 251) and as a preservative agent.

  • Synonyms: Nitric acid sodium salt
  • Molecular Formula: NaNO3
  • Molar Mass: 84.99g/mol
  • CAS: 7631-99-4
  • EIN ECS No.: 231-554-3

Specifications

Melting Point 308°C
CAS 7631-99-4
pH 5.5–8.3 (5%, water)
Quantity 1 kg
Molecular Formula NNaO3
MDL Number MFCD00011119
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
Solubility Information Soluble in water
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
PubChem CID 24268
ChEBI CHEBI:63005
Formula Weight 84.99g/mol
Percent Purity 99.5% min (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate, reagent grade, ACS, ISO
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.