missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, MP Biomedicals

Product Code. 11402766 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402766 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402766 Supplier MP Biomedicals Supplier No. 0521806680

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7631-95-0
Quantity 100 g
Molecular Formula MoNa2O4
Synonym sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES [O-][Mo](=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(dioxo)molybdenum
Molecular Weight (g/mol) 205.926
PubChem CID 61424
ChEBI CHEBI:75215
Formula Weight 205.92
Chemical Name or Material Sodium Molybdate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.