missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, Supelco™

Product Code. 15877480
Change view
Click to view available options
Quantity:
50 kg
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877480 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877480 Supplier Supelco Supplier No. 1065249050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 687°C (Elimination of Water of Crystallisation)
CAS 10102-40-6
Quantity 50 kg
MDL Number MFCD00149170
Synonym Sodium molybdate dihydrate, Sodium molybdate dihydrate
Molecular Weight (g/mol) 241.95
Grade Research
Density 2.71 g/cm3 at 22°C (for the Dihydrate)
Chemical Name or Material Sodium Molybdate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.