missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, Supelco™

Product Code. 15874840
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874840 1 kg 1kg
15864840 100 g 100g
15857480 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15874840 Supplier Supelco Supplier No. 1065211000
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™, ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6

Specifications

Melting Point 687°C (Elimination of water of crystallisation)
Quantity 1 kg
MDL Number MFCD00149170
Synonym Sodium molybdate dihydrate, Sodium molybdate dihydrate
Molecular Weight (g/mol) 241.95
Grade Research
Density 2.71 g/cm3 at 22°C (for the dihydrate)
Chemical Name or Material Sodium Molybdate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.