missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, EMPLURA™, MilliporeSigma™

Product Code. 15867480
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15867480 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15867480 Supplier Supelco Supplier No. 1065241000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity sodium molybdate dihydrate for use in technical applications.

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Specifica

Melting Point 687°C (Elimination of water of crystallization)
CAS 10102-40-6
Color White
pH 9 to 10 (840g/L, H2O, 20°C)
Physical Form Crystals
Quantity 1 kg
Molecular Formula H4MoNa2O6
MDL Number MFCD00149170
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
Solubility Information 840g/L
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
Molecular Weight (g/mol) 241.96
PubChem CID 16211258
ChEBI CHEBI:75213
Formula Weight 241.95
Percent Purity 98.0 to 103.0%
Grade Not Available
Packaging Plastic Bottle
Density 2.71 g/3 (22 °C) (for the dihydrate)
Chemical Name or Material Sodium Molybdate Dihydrate
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.