missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metaperiodate, Supelco™

Product Code. 15877500
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877500 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877500 Supplier Supelco Supplier No. 1065971000
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 270°C (Decomposes)
CAS 7790-28-5
Quantity 1 kg
MDL Number MFCD00003534
Synonym Sodium metaperiodate, Sodium periodate, Sodium metaperiodate
Molecular Weight (g/mol) 213.89
Grade Research
Density 3.87 g/cm3 at 20°C
Chemical Name or Material Sodium Metaperiodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.