missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™
Specifications
Specifications
| Melting Point | 270°C (decomposes) |
| pH | 5.2 (50 g/L, H2O, 20°C) |
| Physical Form | Crystals |
| Quantity | 250 g |
| Assay Percent Range | Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3% |
| Solubility Information | 91 g/L |
| Percent Purity | ≥99.0% |
| Grade | ACS |
| Packaging | Plastic Bottle |
| Density | 3.87 g/cm3 (at 20°C); Bulk Density: 2900 kg/m3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?