missing translation for 'onlineSavingsMsg'
Learn More

Sodium lactate, 50% w/w in water, Pharmpur, Scharlab

Product Code. 11358544
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358544 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11358544 Supplier Scharlab Supplier No. SO04601000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 867-56-1
Quantity 1 L
Molecular Formula C3H5NaO3
InChI Key NGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES [Na+].CC(O)C([O-])=O
IUPAC Name sodium 2-hydroxypropanoate
Molecular Weight (g/mol) 112.06
Grade Pharmpur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.