missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium ionophore III, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
Sodium ionophore III (ETH2120) is a Na+ ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc.
Specifications
Specifications
| Chemical Name or Material | Sodium ionophore III |
| CAS | 81686-22-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H52N2O4 |
| Quantity | 50 mg |
| Synonym | ETH2120 |
| Solubility Information | DMSO : 2 mg/mL (3.62 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (1.81 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(N(C1CCCCC1)C2CCCCC2)COC3=CC=CC=C3OCC(N(C4CCCCC4)C5CCCCC5)=O |
| Molecular Weight (g/mol) | 552.79 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction