missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, Supelco™

Product Code. 15874620
Change view
Click to view available options
Quantity:
2.5 L
25 L
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874620 2.5 L 2.5 L
15847370 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15874620 Supplier Supelco Supplier No. 1056142500
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

6 to 14% Active chlorine EMPLURA™

(6 to 14% active chlorine) EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9

Specifications

Boiling Point 102°C/1013 hPa
Melting Point -20°C
Molecular Formula NaClO
MDL Number MFCD00011120
Density 1.22 to 1.25 g/cm3 at 20°C
Synonym Sodium hypochlorite solution, Soda bleaching lye, Eau de Labarraque, Sodium hypochlorite solution
Molecular Weight (g/mol) 74.44
Grade Reagent
pH 12 to 13 (20°C in H2O)
Quantity 2.5 L
Chemical Name or Material Sodium Hypochlorite Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.