missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, TRC

Product Code. 30008118
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008118 5 g 5g
30003535 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008118 Supplier Toronto Research Chemicals Supplier No. TRCS6240975G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula NaOH
Molecular Weight (g/mol) 40
Synonym Sodium hydroxide, Sodium hydroxide (Na(OH)), Sodium hydroxide (8CI), Aetznatron, Ascarite, BO-200B, Caustic Solution 70TW, Caustic soda, Collo-Grillrein, Collo-Tapetta, DL α, E 33, E 33 (degreasing agent), FC 4370, GR, GR (alkali reagent), MO 67, NSC 135799, Promoter 3308B, Reasol, Soda, caustic, StamperPrep, Tosoh Pearl, White caustic, ma-D 331
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

InChI Formula InChI=1S/Na.H2O/h;1H2/q+1;/p-1
Quantity 5 g
Formula Weight 39.9925
Chemical Name or Material Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.