missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, Supelco™

Product Code. 15873881
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
10 L
25 L
Unit Size:
1 L
10 L
2.5 L
25 L
4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15873881 1 L 1 L
15895230 2.5 L 2.5 L
15805240 4 L 4 L
15815240 10 L 10 L
15867710 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15873881 Supplier Supelco Supplier No. 1091371000
  • ONLINE EXCLUSIVE PRICE
    35.00 EUR
    22.64 EUR / 1 L
    Save 12.36 EUR
    35% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C(NaOH) = 1 mol/L (1 N), Titripur™, reag. Ph. Eur., reag. USP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2

Specifications

Molecular Formula NaOH
MDL Number MFCD00003548
Density 1.04 g/cm3 at 20°C
Quantity 1 L
Molecular Weight (g/mol) 40
Grade Not Available
Chemical Name or Material Sodium Hydroxide Solution
compliance-icons
Product Identifier
  • Sodium hydroxide solution c(NaOH) = 1 mol/l (1 N) Titripur Reag. Ph Eur,Reag. USP
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.