Learn More
Sodium Hydroxide Solution, 1M, Chem-Lab
Product Code. 11913233
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11913233 1 L Plastic Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11913233 Supplier Chem-Lab Supplier No. CL05.1420.1000
  • ONLINE EXCLUSIVE PRICE
    18.30 EUR
    13.60 EUR / 1 L
    Save 4.70 EUR
    25% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
pH >13
Color Colorless
Boiling Point 100°C
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 1M
Density 1.04g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Water Soluble
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Concentration or Composition (by Analyte or Components) 40g NaOH per 1L H2O
Normality 1 N (±0.002, 20°C)
Odor Odorless
Packaging Plastic Bottle
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 1 mol/l
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hydroxide, pellets a.r.