Learn More
WTW™ Sodium Hydroxide Pellets
Product Code. 11745104
Change view
Click to view available options
Type:
Sodium Hydroxide Pellets
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11745104 Sodium Hydroxide Pellets 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11745104 Supplier WTW™ Supplier No. 209140
 Restricted Item
In Stock
Used as a CO2 absorber.
  • Pure sodium hydroxide
  • 2 bottles of 50g each
TRUSTED_SUSTAINABILITY
Specifications
Type Sodium Hydroxide Pellets
For Use With (Application) Used as CO2 absorber
Includes 2 bottles of 50g each
compliance-icons
Product Identifier
  • NHP 600
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide