missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fusidate, Mikromol™
Description
Sodium Fusidate
Specifications
Specifications
| Chemical Name or Material | Fusidic Acid Sodium Salt |
| CAS | 751-94-0 |
| Analyte or Component Names | Fusidic Acid Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C31 H47 O6 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Sodium Fusidate, 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, sodium salt (1:1), (3alpha,4alpha,8alpha,9beta,11alpha,13alpha,14beta,16beta,17Z)-, 29-Nor-8alpha,9beta,13alpha,14beta-dammara-17(20),24-dien-21-oic acid, 3alpha,11alpha,16beta-trihydroxy-, 16-acetate, monosodium salt, (Z)- (8CI), 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, monosodium salt, (3alpha,4alpha,8alpha,9beta,11alpha,13alpha,14beta,16beta,17Z)- (9CI), Fucidin, Fucidina, Fucidine, Fusidate sodium, Fusidic acid sodium salt, Fusidin, SQ 16360, Sodium fucidate, Sodium fusidate, Sodium fusidin, ZN 6-Na, Sodium ent-(17Z)-16alpha-(acetyloxy)-3beta,11beta-dihydroxy-4beta,8,14-trimethyl-18-nor-5beta,10alpha-cholesta-17(20),24-dien-21-oate |
| InChI Formula | InChI=1S/C31H48O6.Na/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);/q;+1/p-1/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1 |
| SMILES | [Na+].C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)[O-])\[C@H](C[C@]34C)OC(=O)C |
| IUPAC Name | sodium;(2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?