missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, Supelco™

Product Code. 15897460
Change view
Click to view available options
Quantity:
250 g
1 kg
50 kg
Unit Size:
1kg
250g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897460 50 kg 50kg
15874820 250 g 250g
15884820 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15897460 Supplier Supelco Supplier No. 1064499050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For analysis EMSURE™ Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4

Specifications

Melting Point 996°C
Quantity 50 kg
MDL Number MFCD00003524
Synonym Sodium fluoride, Sodium fluoride
Molecular Weight (g/mol) 41.99
Grade Research
Density 2.8 g/cm3 at 20°C
Chemical Name or Material Sodium Fluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.