missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium fluoride, Purum p.a., ≥98.0% (F), Solstice
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| CAS | 7681-49-4 |
| Quantity | 25 kg |
| Molecular Formula | FNa |
| MDL Number | MFCD00003524 |
| InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
| SMILES | [F-].[Na+] |
| IUPAC Name | sodium fluoride |
| Molecular Weight (g/mol) | 41.99 |
| Chemical Name or Material | Sodium fluoride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?