missing translation for 'onlineSavingsMsg'
Learn More

Sodium Disulfite, Supelco™

Product Code. 15897480
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
50 kg
Unit Size:
100g
1kg
500g
50kg
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897480 5 kg 5kg
15814850 100 g 100g
15824850 500 g 500g
15834850 1 kg 1kg
15807490 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15897480 Supplier Supelco Supplier No. 1065285000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium metabisulfite for analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4

Specifications

Melting Point 150°C (decomposition)
Quantity 5 kg
MDL Number MFCD00167602
Synonym Sodium disulfite, Sodium metabisulfite, Sodium pyrosulfite, Disulfite, Pyrosulfite, Sodium disulfite
Molecular Weight (g/mol) 190.11
Grade Research
Density 2.36 g/cm3 at 20°C
Chemical Name or Material Sodium Disulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.