Learn More
Sodium Deuteroxide (40% w/w in D2O), TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Deuteroxide |
| CAS | 14014-06-3 |
| Quantity | 5 g |
| Molecular Formula | DNaO |
| Synonym | Deuterated Sodium Hydroxide, Sodium Deuteroxide, Sodium Deuteroxide (NaOD), Sodium Hydroxide-d |
| InChI Formula | InChI=1S/Na.H2O/h;1H2/q+1;/p-1/i/hD |
| SMILES | [Na+].[2H][O-] |
| Molecular Weight (g/mol) | 41 |
| Formula Weight | 40.9988 |
| Concentration or Composition (by Analyte or Components) | 40 % |