Learn More
Sodium Cyanoborohydride, Sigma-Aldrich™
Product Code. 15896550
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896550 25 g 25g
15886550 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896550 Supplier Sigma Aldrich Supplier No. 8180530025
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25895-60-7
Specifications
Melting Point 241°C to 243°C
Quantity 25 g
Density 1.12 g/cm3 at 28°C
MDL Number MFCD00003516
Synonym Sodium cyanoborohydride, Sodium cyanotrihydridoborate, Sodium cyanoborohydride
Molecular Weight (g/mol) 62.84
Grade Research
Chemical Name or Material Sodium Cyanoborohydride