missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium citrate, dihydrate, Supelco™
Specifications
Specifications
| Chemical Name or Material | Sodium citrate, dihydrate |
| CAS | 6132-04-3 |
| Melting Point | 150°C (anhydrous substance), >300°C (lit.) |
| Molecular Formula | HOC(COONa)(CH2COONa)2 · 2H2O |
| Quantity | 25 kg |
| Synonym | Sodium citrate tribasic dihydrate, Trisodium Citrate, Sodium Citrate, Dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate, Sodium citrate tribasic dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate |
| Molecular Weight (g/mol) | 294.1 |
| Grade | Not Available |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?