missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, TRC

Product Code. 30136926
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136926 250 g 250g
30134813 100 g 100g
30008230 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136926 Supplier Toronto Research Chemicals Supplier No. TRCS615180250G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula Cl Na
Molecular Weight (g/mol) 58.44
Synonym Sodium chloride, NaCl
IUPAC Name chlorosodium
SMILES [Na]Cl

Specifications

Quantity 250 g
InChI Formula InChI=1S/ClH.Na/h1H;/q;+1/p-1
Formula Weight 57.9586
Chemical Name or Material Sodium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.