Learn More
Sodium Butyrate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 156-54-7 |
| Melting Point | 244°C |
| Density | 1.324 g/cm3 at 30°C |
| MDL Number | MFCD00002816 |
| Quantity | 100 g |
| Synonym | Sodium butyrate, Butyric acid sodium salt, Sodium butyrate |
| Molecular Weight (g/mol) | 110.09 |
| Grade | Research |