missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, Pharmpur, Scharlab

Product Code. 11348484
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348484 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11348484 Supplier Scharlab Supplier No. SO01701000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7647-15-6
Quantity 1 kg
Molecular Formula BrNa
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES [Na+].[Br-]
IUPAC Name sodium bromide
Molecular Weight (g/mol) 102.89
Grade Pharmpur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.