missing translation for 'onlineSavingsMsg'
Learn More

Sodium Aminosalicylate Dihydrate, Mikromol™

Product Code. 17951076
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951076 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951076 Supplier Mikromol Supplier No. MM1645.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Aminosalicylate Dihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium aminosalicylate dihydrate
CAS 6018-19-5
Analyte or Component Names Sodium aminosalicylate dihydrate
Shipping Condition Room Temperature
Molecular Formula C7 H6 N O3 . Na . 2 H2 O
Certifications/Compliance ISO 17025
Synonym Sodium aminosalicylate dihydrate, Sodium 4-amino-2-hydroxybenzoate dihydrate, Benzoic acid, 4-amino-2-hydroxy-, monosodium salt, dihydrate, Salicylic acid, 4-amino-, monosodium salt, dihydrate, Monosodium p-aminosalicylate dihydrate, Sodium 4-amino-2-hydroxybenzoate dihydrate, Sodium 4-aminosalicylate dihydrate, Sodium p-aminosalicylate dihydrate, p-Aminosalicylic acid sodium salt dihydrate, Benzoic acid, 4-amino-2-hydroxy-, sodium salt, hydrate (1:1:2), Mesalazine Impurity E as Sodium Salt Dihydrate
InChI Formula InChI=1S/C7H7NO3.Na.2H2O/c8-4-1-2-5(7(10)11)6(9)3-4;;;/h1-3,9H,8H2,(H,10,11);;2*1H2/q;+1;;/p-1
SMILES O.O.[Na+].Nc1ccc(C(=O)[O-])c(O)c1
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
Molecular Weight (g/mol) 211.15
Quantity 250 mg
Formula Weight 211.0457 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.