missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-(2-Ethylhexyl) 2-Sulfobutanedioate, TRC

Product Code. 17973175
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973175 50 mg 50mg
17933805 25 mg 25mg
17941895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973175 Supplier Toronto Research Chemicals Supplier No. TRCS62531550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115960-17-3
Molecular Formula C12H21NaO7S
Molecular Weight (g/mol) 332.35
Synonym Docusate Sodium Impurity, 4-(2-Ethylhexyl)ester-2-sulfo-butanedioic Acid Sodium Salt;
IUPAC Name sodium;4-(2-ethylhexoxy)-1-hydroxy-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)O)S(=O)(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H22O7S.Na/c1-3-5-6-9(4-2)8-19-11(13)7-10(12(14)15)20(16,17)18;/h9-10H,3-8H2,1-2H3,(H,14,15)(H,16,17,18);/q;+1/p-1
Formula Weight 332.0906
Chemical Name or Material Sodium 4-(2-Ethylhexyl) 2-Sulfobutanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.