missing translation for 'onlineSavingsMsg'
Learn More

Sodium-2,4-Dinitro Benzene Sulfonate, MP Biomedicals™

Product Code. 11493196 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493196 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493196 Supplier MP Biomedicals Supplier No. 0522224725

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives. MP Biomedicals' Sodium-2,4-Dinitro Benzene Sulfonate is an organosulfur in the Rare Chemical Library.

Specifications

CAS 885-62-1
Melting Point >350°C
Molecular Formula C6H3N2NaO7S
MDL Number MFCD00007471,MFCD10567393
Quantity 25 g
Synonym sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate
InChI Key GSBYVRKLPCSLNV-UHFFFAOYSA-M
SMILES [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O
IUPAC Name sodium 2,4-dinitrobenzene-1-sulfonate
Molecular Weight (g/mol) 270.15
PubChem CID 70700115
Formula Weight 270.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.