Learn More
BRAND™ Soda Lime Glass Petri Dishes
Product Code. 10315571 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
40 mm
60 mm
80 mm
100 mm
150 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Unit Size
10315571 80 mm 15 mm Pack of 10
10624552 100 mm 15 mm Pack of 10
10400873 150 mm 25 mm Pack of 10
10019880 40 mm 12 mm Pack of 10
10450293 60 mm 15 mm Pack of 10
10039890 100 mm 20 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10315571 Supplier BRAND™ Supplier No. 455732
  • ONLINE EXCLUSIVE PRICE
    40.81 EUR
    33.53 EUR / Pack of 10
    Save 7.28 EUR
    17% Off
Estimated Shipment: 22-12-2026
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
  • Dish and lid are flat inside and outside
  • Free from bubbles and streaks
  • Cut edges are fire-polished
  • Suitable for short-term chemical exposure
TRUSTED_SUSTAINABILITY
Specifications
Material Soda Lime Glass
Type Petri Dish
Edges Fire-polished cut edges
Height (Metric) 15 mm
Diameter (Metric) 80 mm