Learn More
Soda Glass Capillary Tube
Product Code. 12877675
Change view
Click to view available options
Length (Metric) Tube:
75 mm
100 mm
Product Type:
Capillary Tube
Capillary Tube, Both Ends Sealed
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tube Product Type Unit Size
12877675 75 mm Capillary Tube Pack of 100
12807685 100 mm Capillary Tube Pack of 100
12897675 100 mm Capillary Tube, Both Ends Sealed Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12877675 Supplier S Murray SAMCO Supplier No. G119/01
  • ONLINE EXCLUSIVE PRICE
    7.30 EUR
    4.17 EUR / Pack of 100
    Save 3.13 EUR
    42% Off
Estimated Shipment: 22-12-2026
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100
Specifications
Product Type Capillary Tube
Length (Metric) Tube 75 mm