missing translation for 'onlineSavingsMsg'
Learn More

SNX18 Antibody - BSA Free, Novus Biologicals™

Product Code. p-200062230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623580 0.02 mL 0.02mL
18693480 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18623580 Supplier Novus Biologicals Supplier No. NBP2934490.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNX18 Polyclonal antibody specifically detects SNX18 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias FLJ11997, FLJ61062, MGC150827, MGC150829, SH3 and PX domain-containing protein 3B, SH3PX2, SH3PXD3BFLJ32560, SNAG1, sorting nexin 18, sorting nexin associated golgi protein 1, sorting nexin-18, Sorting nexin-associated Golgi protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 445-624 of human SNX18 (NP_001096045.1). LNHTANEFARKQVTGFKKEYQKVGQSFRGLSQAFELDQQAFSVGLNQAIAFTGDAYDAIGELFAEQPRQDLDPVMDLLALYQGHLANFPDIIHVQKGALTKVKESRRHVEEGKMEVQKADGIQDRCNTISFATLAEIHHFHQIRVRDFKSQMQHFLQQQIIFFQKVTQKLEEALHKYDSV
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 112574
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.