missing translation for 'onlineSavingsMsg'
Learn More

Sn @ 20μg/mL; Mo, Si @ 100μg/mL in 3% HNO3 (v/v)/tr HF, VHG™

Product Code. 17948189
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948189 100 mL 100mL
17908859 250 mL 250mL
17919409 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17948189 Supplier VHG Labs Supplier No. VHG1206100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sn @ 20μg/mL; Mo, Si @ 100μg/mL in 3% HNO3 (v/v)/tr HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tin/Molybdenum/Silicon
CAS 7440-31-5, 7439-98-7, 7440-21-3
Analyte or Component Names Tin/Molybdenum/Silicon
Shipping Condition Room Temperature
Solvent or Matrix 3% HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Tin 20 μg/mL/Molybdenum 100 μg/mL/Silicon 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.