Learn More
Thermo Scientific™ SMART Digest™ RNase Kits
Product Code. 17716789 Shop All Thermo Scientific Products
Change view
Click to view available options
Enzyme:
RNase A
RNase T1
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Enzyme Unit Size
17716789 RNase T1 1 set
17726789 RNase A 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17716789 Supplier Thermo Scientific™ Supplier No. 60120101
 Restricted Item
Estimated Shipment: 07-10-2026
Use SMART Digest™ RNase Kits in your oligonucleotide mapping and quantitation workflows to obtain high quality analytical results.
Obtain high-quality analytical results from mRNA digestions using Thermo Scientific™ SMART Digest™ RNase Kits. A significant advance in sample preparation for oligonucleotide mapping and quantitation, these kits provide fast and simple mRNA digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that is compatible with automation.
These RNase kits use optimized, heat stable enzymes on magnetic beads, allowing for control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.
Easy and Simple
  • A simple process and control of digestion time ensure required fragment length for comprehensive mRNA sequencing by HRAM-MS
  • Easily automate the entire protocol for high-throughput processing
  • Single nuclease digestion allows full characterization of mRNA
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher data confidence
  • No nuclease contamination of the analytical column
RNase kits are available with with two endoribonuclease enzyme specificities:
  • RNase T1 degrades single-stranded RNA at G residues
  • RNase A degrades single-stranded RNA at C and U residues
TRUSTED_SUSTAINABILITY
Specifications
Cleavage Degrades single-stranded RNA at G residues
Enzyme RNase T1
Format Bulk Magnetic SMART Digest Resin
For Use With (Application) Oligonucleotide Mapping
Product Line SMART Digest
Type RNA Digestion
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Buffer