Learn More
Labconco™ Small Tray: FreeZone™ Stoppering or Bulk Tray Dryer Accessory
Product Code. 15632127 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone™ Tray Dryer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15632127 FreeZone™ Tray Dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15632127 Supplier Labconco™ Supplier No. 7377900
  • ONLINE EXCLUSIVE PRICE
    132.00 EUR
    130.56 EUR / Pack of 1
    Save 1.44 EUR
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-12-2026
Small Tray designed for use with the FreeZone™ Tray Dryer.
Small Tray
  • Stainless steel tray to contain liquids or bulk items for freeze drying
  • Dimensions fit the FreeZone Tray Dryer perfectly, 10.3 in. w x 11.6 in. d x 0.9 in. h
  • Makes sample loading and unloading as well was cleaning easy when using the Tray Dryer (sold separately)
Specifications
No. of Shelves 0
Material Stainless Steel (304)
For Use With (Equipment) FreeZone™ Tray Dryer
Includes Stainless steel tray
Width (English) 10.3 in.
Height (English) 0.9 in.
Length (English) 11.6 in.
Shipping Weight (English) 2.5 lb.