Description
The Micronova SlideMop™ Flat Head Mop Frame and Covers is designed for use in the traditional 'dip and wring' method or as a 'saturation' method where liquid disposal and/or space saving is a factor.
- Collapsible flat head mop frame and polyester covers
- Lightweight, economical design for use throughout the facility
- Quilted polyester mop covers with polyester pad interiors for disinfectant dispersal
- Choice of Stainless Steel or Plastic Frame
- Swivel joint ensures correct lift and pull or figure 8 cleaning techniques
- For use with standard plastic or aluminum threaded handles
- Can be used with SSU Series Stainless Steel Handle with an ACME adapter
- Use with W-94 or W-92 wringer
- Manufactured for use in Controlled Environments, ISO 5 thru 9
- Available gamma irradiated: 25 kGy minimum dose
- Good chemical resistance
- Autoclavable
Specifications
Specifications
| Product Type | Flat Head Mop Cover |
| Color | White |
| Length (English) | 16 in. |
| Sterility | Non-sterile |
| Length (Metric) | 41 cm |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Care Instructions | Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C. |
| For Use With (Equipment) | Buckets and Wringer |
| For Use With (Application) | Cleaning and Sanitization of Cleanroom Surfaces |
| Show More |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?