Learn More
Thermo Scientific™ SL 8 Small Benchtop Centrifuge Series
Product Code. 15132559 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
230 V 50/60 Hz
Refrigerated:
No
Yes
Description:
SL 8 Small Benchtop Centrifuge
SL 8R Small Benchtop Centrifuge
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Refrigerated Description Unit Size
15132559 230 V 50/60 Hz No SL 8 Small Benchtop Centrifuge Each
15221026 230 V 50/60 Hz Yes SL 8R Small Benchtop Centrifuge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15132559 Supplier Thermo Scientific™ Supplier No. 75007221
  • ONLINE EXCLUSIVE PRICE
    3125.00 EUR
    3060.00 EUR / Pack of 1
    Save 65.00 EUR
    2% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform high-capacity work in a compact design with a smart, simple interface and the flexibility to support both clinical and research applications.
Perform high-capacity work in a compact design. The Thermo Scientific™ SL 8 Small Benchtop Centrifuge, available in both ventilated and refrigerated models, features innovative technologies with a smart, simple interface and the flexibility to meet your clinical and research processes and protocols.
Key Innovation:
  • Secure, push-button Thermo Scientific™ Auto-Lock™ rotor exchange in as little as 3 seconds delivers trouble-free rotor installation and removal and clear chamber access for cleaning convenience
  • Biocontainment sealing options, including certified† Thermo Scientific™ ClickSeal™ lids for glove-friendly, one-handed operation
  • Maximized swing-out capacity up to 8 x 50mL conical tubes, 24 x 5/7mL blood tubes, microplates and microtubes, all in one versatile centrifuge
  • Fixed angle flexibility to spin 50mL conical tubes at over 12,000 x g or a wide range of microtubes at speeds up to 30,279 x g
Clinical Applications:
  • Runs up to 24 x 5/7mL blood tubes at a time in swinging bucket configuration
  • Expanded programming options with alphanumeric program naming for pre-saved protocols
  • Auto-Lock rotor exchange simplifies cleaning with full chamber access
  • Conforms to the latest clinical and safety standards
Research Applications:
  • 8 x 50mL swinging bucket rotor capacity, 4 microplates, or 30 filtration columns for research needs
  • Auto-Lock rotor exchange for application flexibility - go from 50mL tube to a microplate to microtubes effortlessly
This Centrifuge also features:
  • Ensure a peaceful laboratory with low noise levels, including just 52 dBA at performance of 17,850 rpm/ 30,279 x g (with SL 8R centrifuge)
  • One-touch operation with pre-saved protocols glove- and detergent-friendly interface
  • Highly visible backlit display for easy reading of parameters across the lab
  • Multilingual instructions- English, Dutch, French, German, Italian, Russian and Spanish - on programming, run conditions, alerts and service messages conforms to the latest clinical and safety standards, such as UL, CE and IVD
† Biocontainment certification by the Public Health England, Porton Down, UK
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 x 145 mL (TX-150), 6 x 50 mL (HIGHConic III)
Max. Speed 4,500 rpm (TX-150 rotor), 16,000 rpm (MicroClick 24 x 2 rotor)
Max. RCF 3,260 x g (TX-150 Rotor); 24,328 x g (MicroClick 24 x 2 Rotor)
Refrigerated No
Height (English) Exterior 12.2 in.
Height (Metric) Exterior 31 cm
Length (English) Exterior 18.9 in.
Length (Metric) Exterior 48 cm
Noise Level <58 dB(A) (with TX-150 rotor); <61 dB(A) (with MicroClick 24 x 2 rotor)
Safety Features Auto-Lock rotor exchange system, SMARTSpin imbalance detection system, certified ClickSeal biocontainment lids
Width (English) Exterior 14.6 in.
Width (Metric) Exterior 37 cm
Wattage 310 w
Certifications/Compliance IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1; UL Listed / CE marked /IVD compliant / Biocontainment certified by the Public Health England, Porton Down, UK
Description SL 8 Small Benchtop Centrifuge
Electrical Requirements 230 V 50/60 Hz
Controller Type Microprocessor
Dimensions (HWD) 12.2 x 14.6 x 18.9 in. (31 x 37 x 48 cm)
Drive System Direct Brushless Induction
Program Storage 4 protocols via direct access key, with naming option and password protection available
Run Time 99 hr. 59 min., continuous
Height (English) Exterior Lid Open 26.8 in.
Profile (Acceleration/Braking) 2 (standard and soft)
Frequency 50/60 Hz
Height (Metric) Exterior Lid Open 67 cm
Rotor TX-150
Voltage 230 V
Model SL 8