Learn More
Notrax Skywalker HD™ ESD Nitrile FR Rubber Tiles with Bubble Pattern
Product Code. 17782148
Change view
Click to view available options
Dimensions (L x W):
91 x 91 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17782148 91 x 91 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17782148 Supplier Notrax Supplier No. 464S0033BL
  • ONLINE EXCLUSIVE PRICE
    326.53 EUR
    265.05 EUR / Pack of 1
    Save 61.48 EUR
    18% Off
Estimated Shipment: 21-12-2026
Heavy duty rubber tiles can be easily assembled for coverage of large areas or individual workstations. Ergonomic benefit derived from a 13 mm thick worker platform with a bubble pattern for maximum fatigue relief in dry industrial environments.
  • Heavy duty rubber tiles of 91 x 91 cm can be easily assembled for coverage of large areas or individual work stations
  • Ergonomic benefit derived from a 13 mm thick worker platform with a bubble pattern for maximum fatigue relief in dry industrial environments
  • A tooled grid of 30 x 30 cm sections on the underside of the mat makes further customization quick and easy
  • Attachable bevelled edges prevent tripping and allow inside and outside corners
  • Slip resistance rating R9 according to DIN 51130
  • Static dissipative natural rubber compound Meets IEC61340-4-1 (category DIF), measured resistance Rg 106 to 109 Ω, Rg 106 to 109 Ω
  • Fire classification Bfl-S1 tested according to EN 13501-1
  • Resistant to most industrial oils and suitable for welding areas
Specifications
Dimensions (L x W) 91 x 91 cm
Color Black
Length (English) 36 in.
Length (Metric) 91 cm
Material ESD and Fire Retardant Nitrile Rubber Compound
Type Industrial and Commercial Matting
Width (Metric) 91 cm